4-Bromo-5-methyl-1H-indazole ≥97%

Supplier: Matrix Scientific

070028-1G
101942-996EA 161.54 USD
101942-996
4-Bromo-5-methyl-1H-indazole ≥97%
4-Bromo-5-methyl-1H-indazole

Hazard: Irritant

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
MDL Number: MFCD11044156
CAS Number: 926922-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR