(S)-1-Boc-3-(hydroxymethyl)piperazine ≥98%

Supplier: Matrix Scientific

Synonyms: (S)-tert-Butyl 3-(hydroxymethyl)piperazine-1-carboxylate

070017-5G
101942-950EA 3515.38 USD
101942-950
(S)-1-Boc-3-(hydroxymethyl)piperazine ≥98%
(S)-1-Boc-3-(hydroxymethyl)piperazine

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₃
MW: 216.2800 g/mol
MDL Number: MFCD05863889
CAS Number: 314741-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR