6-Bromoimidazo[1,2-a]pyridine-8-carboxylic acid ≥95%
Supplier: Matrix Scientific
069984-1G
101942-812EA
2616.92
USD
101942-812
6-Bromoimidazo[1,2-a]pyridine-8-carboxylic acid ≥95%
6-Bromoimidazo[1,2-a]pyridine-8-carboxylic acid
Hazard: Irritant
Formula:
C₈H₅BrN₂O₂ MW: 241.0400 g/mol |
MDL Number:
MFCD08448156 CAS Number: 903129-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...