6-Bromo-2-chloroquinoline ≥95%

Supplier: Matrix Scientific

069977-250MG
101942-780EA 783.15 USD
101942-780
6-Bromo-2-chloroquinoline ≥95%
6-Bromo-2-chloroquinoline

Hazard: Irritant

Formula: C₉H₅BrClN
MW: 242.50 g/mol
MDL Number: MFCD04115272
CAS Number: 1810-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR