3-Bromoisoquinoline ≥97%

Supplier: Matrix Scientific

069897-1G
101942-444EA 112.7 USD
101942-444
3-Bromoisoquinoline ≥97%
3-Bromoisoquinoline

Hazard: Irritant

Formula: C₉H₆BrN
MW: 208.0600 g/mol
MDL Number: MFCD00234479
CAS Number: 34784-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR