6-Bromoisoquinoline ≥97%

Supplier: Matrix Scientific

069896-1G
101942-438EA 15.38 USD
101942-438
6-Bromoisoquinoline ≥97%
6-Bromoisoquinoline

Hazard: Irritant

Formula: C₉H₆BrN
MDL Number: MFCD04973299
CAS Number: 34784-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR