7-Isoquinolinol ≥97%
Supplier: Matrix Scientific
Synonyms:
7-Hydroxyisoquinoline
, Isoquinolin-7-ol
069894-1G
101942-430EA
640
USD
101942-430
7-Isoquinolinol ≥97%
7-Isoquinolinol
Hazard: Irritant
Formula:
C₉H₇NO MW: 145.16 g/mol Melting Pt: 226…228 °C |
MDL Number:
MFCD00456131 CAS Number: 7651-83-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...