2-Methyl-2H-indazole-4-boronic acid pinacol ester ≥96%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole

069864-1G
101942-290EA 769.23 USD
101942-290
2-Methyl-2H-indazole-4-boronic acid pinacol ester ≥96%
2-Methyl-2H-indazole-4-boronic acid pinacol ester

Hazard: Irritant

Formula: C₁₄H₁₉BN₂O₂
MW: 258.1239 g/mol
MDL Number: MFCD11109437
CAS Number: 885698-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR