2,5-Dichloropyrazine ≥97%

Supplier: Matrix Scientific

069847-5G
101942-216EA 135.7 USD
101942-216
2,5-Dichloropyrazine ≥97%
2,5-Dichloropyrazine

Hazard: Irritant

Formula: C₄H₂Cl₂N₂
MW: 148.9781 g/mol
MDL Number: MFCD04117886
CAS Number: 19745-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR