3-Amino-7-fluoro-1-methyl-1H-indazole ≥98%

Supplier: Matrix Scientific

069828-1G
101942-114EA 1044.62 USD
101942-114
3-Amino-7-fluoro-1-methyl-1H-indazole ≥98%
3-Amino-7-fluoro-1-methyl-1H-indazole

Hazard: Irritant

Formula: C₈H₈FN₃
MDL Number: MFCD11109399
CAS Number: 171809-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR