3-Amino-5-fluoro-1-methyl-1H-indazole ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-1-methyl-1H-indazol-3-amine

069826-1G
101942-102EA 880 USD
101942-102
3-Amino-5-fluoro-1-methyl-1H-indazole ≥98%
3-Amino-5-fluoro-1-methyl-1H-indazole

Hazard: Irritant

Formula: C₈H₈FN₃
MW: 165.1700 g/mol
MDL Number: MFCD09054760
CAS Number: 171809-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR