2,6-Difluoro-3-pyridineboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2,6-Difluoropyridine-3-boronic acid

069823-5G
101942-086EA 113.85 USD
101942-086
2,6-Difluoro-3-pyridineboronic acid ≥97%
2,6-Difluoro-3-pyridineboronic acid

Hazard: Irritant/Refrigerate/Stored under Argon

Formula: C₅H₄BF₂NO₂
MW: 158.90 g/mol
Melting Pt: 96…98 °C
MDL Number: MFCD03788245
CAS Number: 136466-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR