4-Methyl-3-pyridineboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Methylpyridin-3-yl)boronic acid4-Methylpyridine-3-boronic acid

069817-5G
101942-058EA 338.46 USD
101942-058
4-Methyl-3-pyridineboronic acid ≥97%
4-Methyl-3-pyridineboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₆H₈BNO₂
MW: 136.94 g/mol
MDL Number: MFCD03788244
CAS Number: 148546-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR