5-(Tributylstannyl)-2-(trifluoromethyl)pyridine ≥96%

Supplier: Matrix Scientific

069814-5G
101942-046EA 1025.8 USD
101942-046
5-(Tributylstannyl)-2-(trifluoromethyl)pyridine ≥96%
5-(Tributylstannyl)-2-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₁₈H₃₀F₃NSn
MDL Number: MFCD11109387
CAS Number: 1204580-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR