2-Amino-5-thiazolecarboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Aminothiazole-5-carboxylic acid

069813-5G
101942-042EA 57.5 USD
101942-042
2-Amino-5-thiazolecarboxylic acid ≥98%
2-Amino-5-thiazolecarboxylic acid

Hazard: Irritant

Formula: C₄H₄N₂O₂S
MW: 144.1500 g/mol
MDL Number: MFCD06203554
CAS Number: 40283-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR