3-Formyl-4-methoxyphenylboronic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Formyl-4-methoxybenzeneboronic acid

069802-5G
101941-998EA 220 USD
101941-998
3-Formyl-4-methoxyphenylboronic acid ≥98%
3-Formyl-4-methoxyphenylboronic acid

Hazard: Irritant

Formula: C₈H₉BO₄
MW: 179.97 g/mol
MDL Number: MFCD02093661
CAS Number: 121124-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR