4-(Hydroxymethyl)phenylboronic acid pinacol ester ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(Hydroxymethyl)phenylboronic acid Pinacol ester(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl alcohol4-(Hydroxymethyl)benzeneboronic acid pinacol ester2-[4-(Hydroxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

069796-5G
101941-974EA 89.7 USD
101941-974
4-(Hydroxymethyl)phenylboronic acid pinacol ester ≥98%
4-(Hydroxymethyl)phenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₃H₁₉BO₃
MW: 234.10 g/mol
MDL Number: MFCD09837617
CAS Number: 302348-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR