1H-Indazole-7-carboxylic acid ≥96%
Supplier: Matrix Scientific
069794-5G
101941-966EA
230.77
USD
101941-966
1H-Indazole-7-carboxylic acid ≥96%
1H-Indazole-7-carboxylic acid
Hazard: Irritant
Formula:
C₈H₆N₂O₂ MW: 162.15 g/mol |
MDL Number:
MFCD06804572 CAS Number: 677304-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...