3-(Chloromethyl)imidazo[1,2-a]pyridine ≥98%

Supplier: Matrix Scientific

069781-5G
101941-906EA 6041.54 USD
101941-906
3-(Chloromethyl)imidazo[1,2-a]pyridine ≥98%
3-(Chloromethyl)imidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₇ClN₂
MDL Number: MFCD06358638
CAS Number: 113855-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR