6-Bromo[1,2,4]triazolo[4,3-a]pyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-[1,2,4]triazolo[4,3-a]pyridine

069763-1G
101941-818EA 640 USD
101941-818
6-Bromo[1,2,4]triazolo[4,3-a]pyridine ≥98%
6-Bromo-[1,2,4]triazolo[4,3-a]pyridine

Hazard: Irritant

Formula: C₆H₄BrN₃
MW: 198.0200 g/mol
MDL Number: MFCD08689506
CAS Number: 108281-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR