8-Bromo-6-methyl[1,2,4]triazolo[1,5-a]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 8-Bromo-6-methyl-[1,2,4]triazolo[1,5-a]pyridine

069762-500MG
101941-814EA 553.85 USD
101941-814
8-Bromo-6-methyl[1,2,4]triazolo[1,5-a]pyridine ≥95%
8-Bromo-6-methyl-[1,2,4]triazolo[1,5-a]pyridine

Hazard: Irritant

Formula: C₇H₆BrN₃
MW: 212.0500 g/mol
MDL Number: MFCD09878402
CAS Number: 957062-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR