Methyl pyrazolo[1,5-a]pyridine-2-carboxylate ≥98%

Supplier: Matrix Scientific

Synonyms: Pyrazolo[1,5-a]pyridine-2-carboxylic acid methyl ester

069759-1G
101941-802EA 584.62 USD
101941-802
Methyl pyrazolo[1,5-a]pyridine-2-carboxylate ≥98%
Methyl pyrazolo[1,5-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₉H₈N₂O₂
MDL Number: MFCD07367039
CAS Number: 151831-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR