Isoquinoline-5-amine ≥98%

Supplier: Matrix Scientific

Synonyms: Isoquinolin-5-amine5-Aminoisoquinoline

069753-5G
101941-772EA 21.54 USD
101941-772
Isoquinoline-5-amine ≥98%
Isoquinoline-5-amine

Hazard: Irritant

Formula: C₉H₈N₂
MW: 144.18 g/mol
MDL Number: MFCD00006907
CAS Number: 1125-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR