4-(Ethylsulfonyl)phenylboronic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(Ethylsulphonyl)benzeneboronic acid

069726-5G 069726-1G
101941-636EA 522.1 USD
101941-636 101941-638
4-(Ethylsulfonyl)phenylboronic acid ≥98%
4-(Ethylsulphonyl)phenylboronic acid

Hazard: Irritant

Formula: C₈H₁₁BO₄S
MW: 214.05
MDL Number: MFCD02093076
CAS Number: 352530-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR