5-Acetyl-2-bromopyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Acetyl-6-bromopyridine

069706-5G
101941-544EA 89.23 USD
101941-544
5-Acetyl-2-bromopyridine ≥98%
5-Acetyl-2-bromopyridine

Hazard: Irritant

Formula: C₇H₆BrNO
MW: 200.03 g/mol
MDL Number: MFCD04974527
CAS Number: 139042-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR