6-Bromoquinoxaline ≥97%
Supplier: Matrix Scientific
069682-5G
101941-422EA
263.08
USD
101941-422
6-Bromoquinoxaline ≥97%
6-Bromoquinoxaline
Hazard: Irritant
Formula:
C₈H₅BrN₂ MW: 209.0427 g/mol |
MDL Number:
MFCD00837757 CAS Number: 50998-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...