Methyl thiazole-4-carboxylate ≥98%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-thiazolecarboxylate
069668-5G
069668-1G
101941-350EA
440
USD
101941-350
101941-348
Methyl thiazole-4-carboxylate ≥98%
Methyl thiazole-4-carboxylate
Hazard: Irritant
Formula:
C₅H₅NO₂S MW: 143.1600 g/mol |
MDL Number:
MFCD06410940 CAS Number: 59418-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...