6-Bromo-1-methyl-1H-indazole ≥98%

Supplier: Matrix Scientific

069658-5G
101941-304EA 146.15 USD
101941-304
6-Bromo-1-methyl-1H-indazole ≥98%
6-Bromo-1-methyl-1H-indazole

Hazard: Irritant

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
MDL Number: MFCD09878569
CAS Number: 590417-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR