4-Benzyloxy-3-methoxyphenylboronic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Benzyloxy-3-methoxybenzeneboronic acid(4-(Benzyloxy)-3-methoxyphenyl)boronic acid

069572-1G 069572-5G
101940-870EA 263.08 USD
101940-870 101940-872
4-Benzyloxy-3-methoxyphenylboronic acid ≥98%
4-Benzyloxy-3-methoxyphenylboronic acid

Hazard: Irritant

Formula: C₁₄H₁₅BO₄
MW: 258.08 g/mol
MDL Number: MFCD09027252
CAS Number: 243990-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR