2-Fluoro-6-(tributylstannyl)pyridine ≥96%
Supplier: Matrix Scientific
069543-1G
069543-25G
069543-5G
101940-708EA
415.38
USD
101940-708
101940-710
101940-712
2-Fluoro-6-(tributylstannyl)pyridine ≥96%
2-Fluoro-6-(tributylstannyl)pyridine
Hazard: Irritant
Formula:
C₁₇H₃₀FNSn |
MDL Number:
MFCD09025793 CAS Number: 1025744-38-0 UN: 2788 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...