3-Bromo-5-(N-boc)aminomethylisoxazole ≥97%

Supplier: Matrix Scientific

Synonyms: 5-(Aminomethyl)-3-bromoisoxazole, 5-BOC protected

069520-5G 069520-500MG 069520-1G
101940-584EA 349.23 USD
101940-584 101940-586 101940-588
3-Bromo-5-(N-boc)aminomethylisoxazole ≥97%
3-Bromo-5-(N-boc)aminomethylisoxazole

Hazard: Irritant

Formula: C₉H₁₃BrN₂O₃
MW: 277.12 g/mol
MDL Number: MFCD06795882
CAS Number: 903131-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR