2-Methoxy-4-(tributylstannyl)thiazole ≥96%
Supplier: Matrix Scientific
069473-500MG
069473-1G
069473-5G
101940-326EA
420
USD
101940-326
101940-328
101940-324
2-Methoxy-4-(tributylstannyl)thiazole ≥96%
2-Methoxy-4-(tributylstannyl)thiazole
Hazard: Irritant
Formula:
C₁₆H₃₁NOSSn MW: 404.20 |
MDL Number:
MFCD07787372 CAS Number: 927391-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...