1-Methyl-2-(tributylstannyl)imidazole ≥80%
Supplier: Matrix Scientific
069465-1G
069465-5G
069465-500MG
101940-276EA
524.62
USD
101940-276
101940-280
101940-278
1-Methyl-2-(tributylstannyl)imidazole ≥80%
1-Methyl-2-(tributylstannyl)imidazole
Hazard: Irritant
Formula:
C₁₆H₃₂N₂Sn MW: 371.15 |
MDL Number:
MFCD01319030 CAS Number: 105494-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...