5-Chloro-2-formylphenylboronic acid ≥97%
Supplier: Matrix Scientific
069461-25G
069461-5G
069461-1G
101940-254EA
349.23
USD
101940-254
101940-256
101940-258
5-Chloro-2-formylphenylboronic acid ≥97%
5-Chloro-2-formylphenylboronic acid
Hazard: Irritant
Formula:
C₇H₆BClO₃ MW: 184.3900 g/mol |
MDL Number:
MFCD03425955 CAS Number: 870238-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...