4-Iodo-1-tritylimidazole ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Iodo-1-trityl-1H-imidazole

069446-5G 069446-1G 069446-25G
101940-166EA 162.15 USD InStock
101940-166 101940-164 101940-168
4-Iodo-1-tritylimidazole ≥95%
4-Iodo-1-tritylimidazole

Purity: 95%
CAS: 96797-15-8
MDL Number: MFCD02179542
Molecular Formula: C22H17IN2
Molecular Weight: 436,3
Hazard: Irritant

Formula: C₂₂H₁₇IN₂
Melting Pt: 224…225 °C
MDL Number: MFCD02179542
CAS Number: 96797-15-8
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR