2-Bromothiazole-4-carbaldehyde ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-4-formylthiazole2-Bromothiazole-4-carboxaldehyde

069436-1G 069436-5G
101940-110EA 449.23 USD
101940-110 101940-108
2-Bromothiazole-4-carbaldehyde ≥98%
2-Bromothiazole-4-carbaldehyde

Hazard: Irritant/Light Sensitive/Refrigerate

Formula: C₄H₂BrNOS
MW: 192.03 g/mol
Melting Pt: 128…130 °C
MDL Number: MFCD04115726
CAS Number: 5198-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR