3-(Boc-amino)phenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (3-((tert-Butoxycarbonyl)amino)phenyl)boronic acid3-(Boc-amino)benzeneboronic acid3-(N-Boc-amino)phenylboronic acid

069415-5G
101939-996EA 135.38 USD
101939-996
3-(Boc-amino)phenylboronic acid ≥97%
3-(Boc-amino)phenylboronic acid

Hazard: Irritant

Formula: C₁₁H₁₆BNO₄
MW: 237.06 g/mol
MDL Number: MFCD03411945
CAS Number: 380430-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR