N-(4-Ethylphenyl)-N'-(6-methyl-4-oxo-1,4-dihydropyrimidin-2-yl)guanidine

Supplier: Matrix Scientific

066088-500MG
101930-956EA 243.08 USD
101930-956
N-(4-Ethylphenyl)-N'-(6-methyl-4-oxo-1,4-dihydropyrimidin-2-yl)guanidine
N-(4-Ethylphenyl)-N'-(6-methyl-4-oxo-1,4-dihydropyrimidin-2-yl)guanidine

Hazard: Irritant

Formula: C₁₄H₁₇N₅O
MDL Number: MFCD19103546
CAS Number: 1306739-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR