N-(4-Fluorophenyl)-2-mercaptoacetamide

Supplier: Matrix Scientific

066014-500MG
101930-808EA 364.62 USD
101930-808
N-(4-Fluorophenyl)-2-mercaptoacetamide
N-(4-Fluorophenyl)-2-mercaptoacetamide

Hazard: Irritant

Formula: C₈H₈FNOS
MDL Number: MFCD00115449
CAS Number: 70453-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR