Methyl (4-amino-1,2,5-oxadiazol-3-yl)carbamate

Supplier: Matrix Scientific

066013-500MG
101930-806EA 484.62 USD
101930-806
Methyl (4-amino-1,2,5-oxadiazol-3-yl)carbamate
Methyl (4-amino-1,2,5-oxadiazol-3-yl)carbamate

Hazard: Irritant

Formula: C₄H₆N₄O₃
MDL Number: MFCD19103534
CAS Number: 1256628-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR