4,6-Dimethyl-N-(octahydro-2H-benzimidazol-2-ylidene)pyrimidin-2-amine

Supplier: Matrix Scientific

066006-500MG
101930-792EA 364.62 USD
101930-792
4,6-Dimethyl-N-(octahydro-2H-benzimidazol-2-ylidene)pyrimidin-2-amine
4,6-Dimethyl-N-(octahydro-2H-benzimidazol-2-ylidene)pyrimidin-2-amine

Hazard: Irritant

Formula: C₁₃H₁₉N₅
MDL Number: MFCD19103531
CAS Number: 575461-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR