1-(2-Fluorophenyl)-4-piperidinone

Supplier: Matrix Scientific

065774-500MG
101930-330EA 146.15 USD
101930-330
1-(2-Fluorophenyl)-4-piperidinone
1-(2-Fluorophenyl)-4-piperidinone

Hazard: Irritant

Formula: C₁₁H₁₂FNO
MDL Number: MFCD05864574
CAS Number: 115012-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR