3-Cyanobenzamide

Supplier: Matrix Scientific

065633-500MG
101930-070EA 181.7 USD
101930-070
3-Cyanobenzamide
3-Cyanobenzamide

Hazard: Irritant

Formula: C₈H₆N₂O
MW: 146.1460 g/mol
Melting Pt: 222 °C
MDL Number: MFCD00017574
CAS Number: 3441-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR