Methyl-3-amino-5-bromobenzoate

Supplier: Matrix Scientific

Synonyms: Methyl 3-amino-5-bromobenzoate

065397-500MG
101929-594EA 61.54 USD
101929-594
Methyl-3-amino-5-bromobenzoate
Methyl-3-amino-5-bromobenzoate

Hazard: Irritant

Formula: C₈H₈BrNO₂
MW: 230.0586 g/mol
MDL Number: MFCD07786958
CAS Number: 706791-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR