3-(4-Methoxyphenyl)thiomorpholine

Supplier: Matrix Scientific

065211-500MG
101929-224EA 484.62 USD
101929-224
3-(4-Methoxyphenyl)thiomorpholine
3-(4-Methoxyphenyl)thiomorpholine

Hazard: Irritant

Formula: C₁₁H₁₅NOS
CAS Number: 887344-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR