5-tert-Butyl-L-glutamate hydrate
Supplier: Matrix Scientific
Synonyms:
L-Glutamic Acid 5-tert-Butyl Ester
, H-Glu(OtBu)-OH
065141-5G
065141-25G
101929-082EA
46.15
USD
101929-082
101929-084
5-tert-Butyl-L-glutamate hydrate
5-tert-Butyl-L-glutamate hydrate
Hazard: Irritant/Refrigerate
Formula:
C₉H₁₇NO₄ MW: 203.24 g/mol |
MDL Number:
MFCD00038580 CAS Number: 2419-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...