(Carbobenzoxy)glycylglycine

Supplier: Matrix Scientific

Synonyms: Z-Gly-Gly-OH2-(2-(((Benzyloxy)carbonyl)amino)acetamido)acetic acid

065135-500MG
101929-070EA 243.08 USD
101929-070
(Carbobenzoxy)glycylglycine
(Carbobenzoxy)glycylglycine

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₅
MW: 266.25 g/mol
MDL Number: MFCD00037783
CAS Number: 2566-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR