3-Fluoro-N-isopropylbenzylamine

Supplier: Matrix Scientific

060744-500MG
101920-600EA 484.62 USD
101920-600
3-Fluoro-N-isopropylbenzylamine
3-Fluoro-N-isopropylbenzylamine

Hazard: Irritant

Formula: C₁₀H₁₄FN
MDL Number: MFCD08060644
CAS Number: 90389-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR