3-(Piperidin-1-ylmethyl)phenylboronic acid hydrochloride

Supplier: Matrix Scientific

060726-500MG
101920-564EA 969.23 USD
101920-564
3-(Piperidin-1-ylmethyl)phenylboronic acid hydrochloride
3-(Piperidin-1-ylmethyl)phenylboronic acid hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉BClNO₂
MDL Number: MFCD10699627
CAS Number: 1072946-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR