3-Amino-N,N-dimethyl-1H-1,2,4-triazole-5-carboxamide hydrochloride

Supplier: Matrix Scientific

060611-500MG
101920-334EA 484.62 USD
101920-334
3-Amino-N,N-dimethyl-1H-1,2,4-triazole-5-carboxamide hydrochloride
3-Amino-N,N-dimethyl-1H-1,2,4-triazole-5-carboxamide hydrochloride

Hazard: Irritant

Formula: C₅H₁₀ClN₅O
MDL Number: MFCD16618456
CAS Number: 1228552-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR